EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
General approach to epipolythiodiketopiperazine alkaloids: total synthesis of (+)-chaetocins A and C and (+)-12,12'-dideoxychetracin A.
PMID 20866039
Abstract
A highly stereoselective and systematic strategy for the introduction of polysulfides in the synthesis of epipolythiodiketopiperazines is described. We report the first total synthesis of dimeric epitri- and epitetrathiodiketopiperazines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9492
Chaetocin from Chaetomium minutum, ≥95% (HPLC)
C30H28N6O6S4