EMAIL THIS PAGE TO A FRIEND
Cellular immunology
Defining the immunological phenotype of Fc receptor-like B (FCRLB) deficient mice: Confounding role of the inhibitory FcγRIIb.
PMID 20869045
Abstract
Fc receptor-like A (FCRLA) and FCRLB have homology to the transmembrane FCRL family members (FCRL 1-6) and to the conventional receptors for the Fc portion of immunoglobulin, but uniquely are cytosolic proteins expressed in B cells. Here we describe the phenotype of Fcrlb-gene targeted mice. B cell development and in vitro responses are normal; however, antibody responses to a T-dependent antigen are elevated. The gene encoding the inhibitory FcγRIIb is located nearby Fcrlb. Although Fcrlb-gene targeting had no effect on the function or basal expression of FcγRIIb, its expression was reduced following activation. This abnormal regulation was due to co-inheritance of Fcgr2b and the mutant Fcrlb allele from the 129 ES cells. A promoter polymorphism in the 129/Sv Fcgr2b allele results in diminished upregulation of FcγRIIb following B cell activation. Thus, we speculate that the enhanced antibody response seen in the FCRLB-deficient mice may be due to the Fcgr2b promoter.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219924
4-Hydroxy-3-nitrophenylacetic acid, 99%
C8H7NO5