EMAIL THIS PAGE TO A FRIEND
Journal of molecular and cellular cardiology
Small heat shock protein 20 (HspB6) in cardiac hypertrophy and failure.
PMID 20869365
Abstract
Hsp20, referred to as HspB6, is constitutively expressed in various tissues. Specifically, HspB6 is most highly expressed in different types of muscle including vascular, airway, colonic, bladder, and uterine smooth muscle; cardiac muscle; and skeletal muscle. It can be phosphorylated at Ser-16 by both cAMP- and cGMP-dependent protein kinases (PKA/PKG). Recently, Hsp20 and its phosphorylation have been implicated in multiple physiological and pathophysiological processes including smooth muscle relaxation, platelet aggregation, exercise training, myocardial infarction, atherosclerosis, insulin resistance and Alzheimer's disease. In the heart, key advances have been made in elucidating the significance of Hsp20 in contractile function and cardioprotection over the last decade. This mini-review highlights exciting findings in animal models and human patients, with special emphasis on the potential salutary effects of Hsp20 in heart disease. This article is part of a special issue entitled "Key Signaling Molecules in Hypertrophy and Heart Failure."
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV48436
Anti-HSPB6 antibody produced in rabbit, IgG fraction of antiserum