EMAIL THIS PAGE TO A FRIEND
Sexually transmitted diseases
The usefulness of toluidine red unheated serum test in the diagnosis of HIV-negative neurosyphilis.
PMID 20871491
Abstract
We retrospectively analyzed the Venereal Disease Research Laboratory test, Treponema pallidum particle agglutination test, and toluidine red unheated serum test (TRUST) in 41 cases of HIV-negative neurosyphilis and 34 non-neurologic syphilitic patient and found that serum-TRUST titers could be the indication of lumbar puncture in syphilitic patients and a reactive cerebrospinal fluid-TRUST is considered diagnostic to neurosyphilis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59659
Toluidine Red, analytical standard
C17H13N3O3
199753
Toluidine Red, Dye content 70 %
C17H13N3O3