EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Ibrolipim increases ABCA1/G1 expression by the LXRα signaling pathway in THP-1 macrophage-derived foam cells.
PMID 20871621
Abstract
To determine the effects and potential mechanisms of ibrolipim on ATP-binding membrane cassette transporter A-1 (ABCA1) and ATP-binding membrane cassette transporter G-1 (ABCG1) expression from human macrophage foam cells, which may play a critical role in atherogenesis. Human THP-1 cells pre-incubated with ox-LDL served as foam cell models. Specific mRNA was quantified using real-time RT-PCR and protein expression using Western blotting. Cellular cholesterol handling was studied using cholesterol efflux experiments and high performance liquid chromatography assays. Ibrolipim 5 and 50 μmol/L significantly increased cholesterol efflux from THP-1 macrophage-derived foam cells to apoA-I or HDL. Moreover, it upregulated the expression of ABCA1 and ABCG1. In addition, LXRα was also upregulated by the ibrolipim treatment. In addition, LXRα small interfering RNA completely abolished the promotion effect that was induced by ibrolipim. Ibrolipim increased ABCA1 and ABCG1 expression and promoted cholesterol efflux, which was mediated by the LXRα signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1540
NO-1886
C19H20BrN2O4P