EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Keratan sulfate suppresses cartilage damage and ameliorates inflammation in an experimental mice arthritis model.
PMID 20875399
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1031
Anti-Aggrecan Antibody, clone, Chemicon®, from rabbit