EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Improved GC-MS procedure for simultaneous measurement of ethylene glycol and glycolic acid.
PMID 20876228
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323594
α-Hydroxyisobutyric acid, 99%
C4H8O3
164976
α-Hydroxyisobutyric acid, 98%
C4H8O3