EMAIL THIS PAGE TO A FRIEND
Diabetes
Intranasal insulin enhances postprandial thermogenesis and lowers postprandial serum insulin levels in healthy men.
PMID 20876713
Abstract
Animal studies indicate a prominent role of brain insulin signaling in the regulation of peripheral energy metabolism. We determined the effect of intranasal insulin, which directly targets the brain, on glucose metabolism and energy expenditure in humans. In a double-blind, placebo-controlled, balanced within-subject comparison, 19 healthy normal-weight men (18-26 years old) were intranasally administered 160 IU human insulin after an overnight fast. Energy expenditure assessed via indirect calorimetry and blood concentrations of glucose, insulin, C-peptide, and free fatty acids (FFAs) were measured before and after insulin administration and the subsequent consumption of a high-calorie liquid meal of 900 kcal. Intranasal insulin, compared with placebo, increased postprandial energy expenditure, i.e., diet-induced thermogenesis, and decreased postprandial concentrations of circulating insulin and C-peptide, whereas postprandial plasma glucose concentrations did not differ from placebo values. Intranasal insulin also induced a transient decrease in prandial serum FFA levels. Enhancing brain insulin signaling by means of intranasal insulin administration enhances the acute thermoregulatory and glucoregulatory response to food intake, suggesting that central nervous insulin contributes to the control of whole-body energy homeostasis in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5523 Insulin from porcine pancreas, powder, ≥27 USP units/mg
C256H381N65O76S6