EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Phenyl ring-substituted lobelane analogs: inhibition of [³H]dopamine uptake at the vesicular monoamine transporter-2.
PMID 20876747
Abstract
Lobeline attenuates the behavioral effects of methamphetamine via inhibition of the vesicular monoamine transporter (VMAT2). To increase selectivity for VMAT2, chemically defunctionalized lobeline analogs, including lobelane, were designed to eliminate nicotinic acetylcholine receptor affinity. The current study evaluated the ability of lobelane analogs to inhibit [³H]dihydrotetrabenazine (DTBZ) binding to VMAT2 and [³H]dopamine (DA) uptake into isolated synaptic vesicles and determined the mechanism of inhibition. Introduction of aromatic substituents in lobelane maintained analog affinity for the [³H]DTBZ binding site on VMAT2 and inhibitory potency in the [³H]DA uptake assay assessing VMAT2 function. The most potent (K(i) = 13-16 nM) analogs in the series included para-methoxyphenyl nor-lobelane (GZ-252B), para-methoxyphenyl lobelane (GZ-252C), and 2,4-dichlorphenyl lobelane (GZ-260C). Affinity of the analogs for the [³H]DTBZ binding site did not correlate with inhibitory potency in the [³H]DA uptake assay. It is noteworthy that the N-benzylindole-, biphenyl-, and indole-bearing meso-analogs 2,6-bis[2-(1-benzyl-1H-indole-3-yl)ethyl]-1-methylpiperidine hemifumarate (AV-1-292C), 2,6-bis(2-(biphenyl-4-yl)ethyl)piperidine hydrochloride (GZ-272B), and 2,6-bis[2-(1H-indole-3-yl)ethyl]-1-methylpiperidine monofumarate (AV-1-294), respectively] inhibited VMAT2 function (K(i) = 73, 127, and 2130 nM, respectively), yet had little to no affinity for the [³H]DTBZ binding site. These results suggest that the analogs interact at an alternate site to DTBZ on VMAT2. Kinetic analyses of [³H]DA uptake revealed a competitive mechanism for 2,6-bis(2-(4-methoxyphenyl)ethyl)piperidine hydrochloride (GZ-252B), 2,6-bis(2-(4-methoxyphenyl)ethyl)-1-methylpiperidine hydrochloride (GZ-252C), 2,6-bis(2-(2,4-dichlorophenyl)ethyl)piperidine hydrochloride (GZ-260C), and GZ-272B. Similar to methamphetamine, these analogs released [³H]DA from the vesicles, but with higher potency. In contrast to methamphetamine, these analogs had higher potency (>100-fold) at VMAT2 than DAT, predicting low abuse liability. Thus, modification of the lobelane molecule affords potent, selective inhibitors of VMAT2 function and reveals two distinct pharmacological targets on VMAT2.