EMAIL THIS PAGE TO A FRIEND
Journal of oleo science
Antioxidant activity of colored rice bran obtained at different milling yields.
PMID 20877150
Abstract
In this study, we investigated the antioxidant components of three types of colored rice bran--forbidden rice, red rice and green rice--obtained from rice in which the pigment layer had been removed at milling yields of 90%-100% and 80%-90%. An evaluation of the effects of 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical scavenging activity revealed that rice bran obtained from forbidden rice at milling yields of 90%-100% and 80%-90% and rice bran obtained from red rice at milling yields of 90%-100% showed favorable antioxidant activity. The antioxidant components were confirmed to be 3,4-dihydroxy methyl benzoate and p-methoxyphenol and they influence the antioxidant activity of the three types of colored rice bran.
Related Materials