EMAIL THIS PAGE TO A FRIEND
Metal ions in life sciences
Alkyl derivatives of bismuth in environmental and biological media.
PMID 20877811
Abstract
Knowledge about methylated species of bismuth in environmental and biological media is very limited. The presence of volatile trimethylbismuthine has been unequivocally detected in landfill and sewage fermentation gases but the trace concentrations of methylated bismuth species reported in a few polluted soils and sediments probably require further confirmation. In contrast to arsenic and antimony, no methylated bismuth species have ever been found in surface waters and biota. Volatile monomethyl-, dimethyl- and trimethylbismuthine have been produced by some anaerobic bacteria and methanogenic archaea in laboratory culture experiments. Bismuth methylation differs significantly from the one of arsenic and antimony because no Bi(V) compound is known to be formed in biological and environmental media. Moreover, alkylbismuth compounds are rather instable due to the easy cleavage of the weak Bi-C bond.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86561354
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF85045631
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF80139438
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF60966366
Bismuth, rod, 70mm, diameter 10.0mm, cast sticks, 100%
Bi
GF89918500
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bi
GF71570550
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Bi
GF58555581
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bi
GF52383506
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Bi
GF31001960
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Bi
GF39813419
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Bi
GF46319340
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Bi
GF19448017
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Bi
GF97656238
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bi
GF75246183
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bi
GF46489135
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF86892441
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF40931546
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF35246831
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF95571995
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF12550935
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF09655263
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF71673060
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF09816307
Bismuth, foil, light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF91742328
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF24083692
Bismuth, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF77293592
Bismuth, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF93912891
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF19216087
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF64303455
Bismuth, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF30816356
Bismuth, foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi