EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Synthesis of the core structure of the lipoteichoic acid of Streptococcus pneumoniae.
PMID 20878800
Abstract
Streptococcus pneumoniae LTA is a highly complex glycophospholipid that consists of nine carbohydrate residues: three glucose, two galactosamine and two 2-acetamino-4-amino-2,4,6-trideoxygalactose (AATDgal) residues that are each differently linked, one ribitol and one diacylated glycerol (DAG) residue. Suitable building blocks for the glucose and the AATDgal residues were designed and their synthesis is described in this paper. These building blocks permitted the successful synthesis of the core structure Glcβ(1-3)AATDgalβ(1-3)Glcα(1-O)DAG in a suitably protected form for further chain extension (1 b, 1 c) and as unprotected glycolipid (1 a) that was employed in biological studies. These studies revealed that 1 a as well as 1 lead to interleukin-8 release, however not via TLR2 or TLR4 as receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5502
Adonitol, ≥99%
C5H12O5
02240
Adonitol, BioXtra, ≥99.0% (HPLC)
C5H12O5