EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of amidomethyltrifluoroborates and their use in cross-coupling reactions.
PMID 20879742
Abstract
Amidomethyltrifluoroborates were successfully synthesized in a one-pot fashion and used in cross-coupling reactions with a wide variety of aryl and heteroaryl chlorides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
207934
Tetrafluoroboric acid solution, 48 wt. % in H2O
HBF4