EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
An efficient method for the selective synthesis of 2-deoxy-2-iodo-glycosides by O-glycosidation of D-glucal using I₂-Cu(OAc)₂.
PMID 20880519
Abstract
An efficient and convenient method for the synthesis of 2-deoxy-2-iodo-O-glycosides from tri-O-acetyl-D-glucal with various alcohols by using I₂-Cu(OAc)₂ is described. The 21 examples of corresponding glycosides were obtained in high yields, with good anomeric selectivity.
Related Materials