EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
A facile chemical approach for preparing a SERS active silver substrate.
PMID 20882223
Abstract
This communication describes a new surface-enhanced Raman scattering (SERS) active silver substrate prepared by iodination of the evaporated silver foil. After iodination, the morphology of the silver substrate undergoes a self-evolution process in which it displays accordingly the UV-vis absorption shift as well as the AFM topological test. Rhodamine 6G (R6G) is used as the probe molecule to evaluate the enhancement efficiency of the silver substrate at different self-evolution time intervals. The SERS intensity of R6G increases up to ∼29-fold and reaches a maximum after the substrate evolved for 24 h. This method is feasible for the production of an efficient SERS silver substrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF90975516
Silver, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95+%
Ag
GF23006797
Silver, foil, 25x25mm, thickness 0.1mm, annealed, 99.95+%
Ag
GF29990661
Silver, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95+%
Ag
GF74445813
Silver, foil, 25x25mm, thickness 0.20mm, as rolled, 99.95+%
Ag
GF70015946
Silver, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95+%
Ag
GF46518903
Silver, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Ag
GF10931947
Silver, foil, 0.5m coil, thickness 0.01mm, 99.95+%
Ag
GF64714215
Silver, foil, 0.5m coil, thickness 0.025mm, annealed, 99.95+%
Ag
GF40017620
Silver, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.95+%
Ag
GF62467386
Silver, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Ag
GF24663012
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Ag
GF51746250
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Ag
GF10306987
Silver, foil, 0.5m coil, thickness 0.05mm, annealed, 99.95+%
Ag
GF40779726
Silver, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Ag
GF83898905
Silver, foil, 0.5m coil, thickness 0.125mm, annealed, 99.95+%
Ag
GF71813785
Silver, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Ag
GF27190762
Silver, foil, 0.5m coil, thickness 0.125mm, coil width 300mm, annealed, 99.95+%
Ag
GF07805695
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Ag
GF23632332
Silver, foil, 0.5m coil, thickness 0.1mm, annealed, 99.95+%
Ag
GF58225518
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Ag
GF65202599
Silver, foil, 10mm disks, thickness 0.0025mm, 99.97%
Ag
GF96071104
Silver, foil, 10mm disks, thickness 0.003mm, 99.9%
Ag
GF78575732
Silver, foil, 10mm disks, thickness 0.004mm, 99.97%
Ag
GF05418312
Silver, foil, 10mm disks, thickness 0.005mm, 99.97%
Ag
GF35518169
Silver, foil, 10mm disks, thickness 0.007mm, 99.97%
Ag
GF25600777
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Ag
GF15732402
Silver, foil, 10mm disks, thickness 0.008mm, 99.97%
Ag
GF15118253
Silver, foil, 10mm disks, thickness 0.009mm, 99.97%
Ag
GF25755217
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Ag
GF92043244
Silver, foil, 10mm disks, thickness 0.015mm, 99.97%
Ag