EMAIL THIS PAGE TO A FRIEND
Comparative Biochemistry and Physiology. Toxicology & Pharmacology : CBP
Genotoxic and cytotoxic effects induced by aluminum in the lymphocytes of the common carp (Cyprinus carpio).
PMID 20883821