EMAIL THIS PAGE TO A FRIEND
Nature protocols
A quantitative high-throughput 96-well plate fluorescence assay for mechanism-based inactivators of cytochromes P450 exemplified using CYP2B6.
PMID 20885377
Abstract
Mechanism-based inactivators such as bergamottin are useful chemical tools for identifying the functions of specific active-site amino acid residues in the reactions catalyzed by cytochromes P450 (CYPs or P450s), which are responsible for the metabolism of a wide variety of drugs and endogenous substrates. In clinical settings, mechanism-based inactivation of P450s involved in xenobiotic metabolism has the potential to lead to adverse drug-drug interactions, and assays to identify and characterize drug candidates as P450 inactivators are important in drug discovery and development. Here we present a quantitative high-throughput protocol for investigating cytochrome P450 mechanism-based inactivators; we use the example of CYP2B6 and bergamottin to illustrate the finer points of this protocol. This protocol details the adaptation of a 7-ethoxytrifluoromethyl coumarin O-deethylation fluorescence activity assay to a 96-well microtiter plate format and uses a plate reader to detect the fluorescence of the product. Compared with previous methods, this protocol requires less P450 and takes significantly less time while greatly increasing throughput. The protocol as written takes ∼2 h to complete. The principles and procedures outlined in this protocol can be easily adapted to other inactivators, P450 isoforms, substrates and plate readers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2803
7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)
C12H9F3O3
01338
Bergamottin, analytical standard
C21H22O4