EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Apatite germanates doped with tungsten: synthesis, structure, and conductivity.
PMID 20886149
Abstract
High oxygen content apatite germanates, La(10)Ge(6-x)W(x)O(27+x), have been prepared by doping on the Ge site with W. In addition to increasing the oxygen content, this doping strategy is shown to result in stabilisation of the hexagonal lattice, and yield high conductivities. Structural studies of La(10)Ge(5.5)W(0.5)O(27.5) show that the interstitial oxygen sites are associated to a different degree with the Ge/WO(4) tetrahedra, leading to five coordinate Ge/W and significant disorder for the oxygen sites associated with these units. Raman spectroscopy studies suggest that in the case of the WO(5) units, the interstitial oxygen is more tightly bonded and therefore not as mobile as in the case of the GeO(5) units, thus not contributing significantly to the conduction process.