EMAIL THIS PAGE TO A FRIEND
Nanoscale
Destabilisation of complex hydrides through size effects.
PMID 20886168
Abstract
Nanoparticles of NaAlH4, LiAlH4 and LiBH4 were prepared by encapsulating their respective hydrides within carbon nanotubes by a wet chemical approach. The resulting confinement had a profound effect on the overall hydrogen storage properties of these hydrides, with NaAlH4 and LiAlH4 releasing hydrogen from room temperature, for example.