EMAIL THIS PAGE TO A FRIEND
Australian dental journal
In vitro study of the inactivation by dentine of some endodontic medicaments and their bases.
PMID 20887518
Abstract
The aim of this study was to investigate the antimicrobial effect of endodontic medicaments and their bases in the presence of dentine powder. The medicaments tested were Ledermix paste, Pulpdent paste, a 50:50 combination of the Pulpdent:Ledermix and their bases. The test organism was Enterococcus faecalis ATCC 29212. The presence or absence of dentine was examined as well as the effect of autoclaving dentine. Serial dilutions of samples at 1 hour, 1 day and 3 days were used for colony counting. The effects of dentine powder on pH for saturated Ca(OH)2 solution and Pulpdent paste at 1 hour and 24 hours were also measured. Pulpdent and the 50:50 combination of Pulpdent:Ledermix completely inhibited the growth of E. faecalis from 1 hour onwards, and these results were not affected by the presence/absence of dentine powder, pre-incubation period, timing of autoclaving, or exposure time. Saturated solutions of Ca(OH)2 are prone to inactivation by dentine powder unlike Pulpdent paste. Ledermix paste took 3 days to exert a significant effect on the growth of E. faecalis. In this laboratory study, both Pulpdent and the 50:50 mixture of Pulpdent with Ledermix were effective medicaments against E. faecalis in the presence of dentine powder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6140
Demeclocycline hydrochloride, powder or crystals
C21H21ClN2O8 · HCl
46161
Demeclocycline hydrochloride hydrate, VETRANAL®, analytical standard
C21H21ClN2O8 · HCl · xH2O