EMAIL THIS PAGE TO A FRIEND
Parasitology international
Multiple resistance to anthelmintics by Haemonchus contortus and Trichostrongylus colubriformis in sheep in Brazil.
PMID 20887800
Abstract
The objective of this study was to determine the level of resistance of Haemonchus contortus and Trichostrongylus colubriformis in sheep to levamisole, albendazole, ivermectin, moxidectin, closantel and trichlorfon. The parasites were isolated from sheep naturally infected by gastrointestinal nematodes and were then kept in monospecifically-infected lambs for production of infective larvae (L3) of both species. Forty-two lambs, at three months of age, were simultaneously artificially infected with 4000 L3 of H. contortus and 4000 L3 of T. colubriformis. The animals were allocated into seven groups with six animals each that received one of the following treatments: Group 1--control, no treatment; Group 2--moxidectin (0.2mg/kg body weight (BW)); Group 3--closantel (10mg/kg BW); Group 4--trichlorfon (100mg/kg BW); Group 5--levamisole phosphate (4.7 mg/kg BW); Group 6--albendazole (5.0mg/kg BW); and Group 7--ivermectin (0.2mg/kg BW). Nematode fecal egg counts (FEC) were carried out on the day of treatment and again at 3, 7, 10 and 14 days post-treatment. On the same occasions, composite fecal cultures were prepared for each group for production of L3, which were identified into genus. The animals were sacrificed for worm counts at 14 days after treatment. The efficacy of each treatment was calculated from the arithmetic mean of the FEC or worm burden of the treated group, compared with the values of the control group. Only trichlorfon and moxidectin treatments resulted in a significant reduction of H. contortus recorded at necropsy (73% and 45% respectively). Moxidectin reduced T. colubriformis worm burdens by 82% and albendazole by 19%. All other anthelmintics resulted in no significant reduction in the numbers of worms found at necropsy. In conclusion, the isolates of H. contortus and T. colubriformis showed multiple resistance to all groups of anthelmintics tested. This is the first report, based on the controlled efficacy test, to show resistance of T. colubriformis to macrocyclic lactones in Brazil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34093
Closantel, PESTANAL®, analytical standard
C22H14Cl2I2N2O2