EMAIL THIS PAGE TO A FRIEND
Life sciences
The possible involvement of dopamine D3 receptors in the regulation of gastric emptying in rats.
PMID 20888837
Abstract
The inhibitory effect of dopamine on gastric motility is thought to be mediated via a decrease in acetylcholine release resulting from stimulation of enteric neuronal dopamine D(2) receptors. The aim of this study was to investigate the possible involvement of the dopamine D(3) receptor in the regulation of gastric motility in rats using selective dopamine D(3) receptor agonists or a dopamine D(3) receptor antagonist. Gastric emptying was assessed using the phenol red method after rats were treated with varying doses of dopamine D(3) receptor agonists or a dopamine D(3) receptor antagonist. S(+)-PD 128,907 (0.01-1 mg/kg, s.c.), a selective dopamine D(3) receptor agonist, dose-dependently delayed gastric emptying in rats. Other dopamine D(3) receptor agonists (i.e., R(+)-7-OH-DPAT [0.03-1 mg/kg, s.c.] and quinpirole [0.01-1 mg/kg, s.c.]) also delayed gastric emptying in rats. Both the selective dopamine D(1) and D(5) receptor agonist SKF-38393 and the selective dopamine D(4) receptor agonist PD 168,077 failed to delay gastric emptying in rats. The selective dopamine D(3) receptor antagonist (+)-S 14297 (10mg/kg, s.c.) partially inhibited the S(+)-PD 128,907-induced delay in gastric emptying. Although an administration of S(+)-PD 128,907 (1-100 μg/kg) into the 4th cerebral ventricle partially and dose-dependently delayed gastric emptying in rats, its administration into the lateral cerebral ventricle did not affect gastric emptying. The results presented here suggest that peripheral dopamine D(2) receptors and, at least in part, dopamine D(3) and central dopamine D(2)/D(3) receptors play an important role in the regulation of gastric motility in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H168
R(+)-7-Hydroxy-DPAT hydrobromide, >98%, solid
C16H25NO · HBr