EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
A method for analysis of dimethyl selenide and dimethyl diselenide by LC-ICP-DRC-MS.
PMID 20890781
Abstract
The aim of this work was to develop a simple and fast high performance liquid chromatography-inductively coupled argon plasma (ICP) mass spectrometry (MS) method capable of separating and detecting the two volatile selenium species dimethyl selenide (DMeSe) and dimethyl diselenide (DMeDSe) in biological samples. Dimethyl selenide and dimethyl diselenide were separated on a short reversed phase column using an eluent containing 40% methanol and detected by dynamic reaction cell ICP-MS monitoring the (80)Se isotope. The limit of detection was 8 nM for both species (corresponding to 0.6 and 1.3 μg Se/L for DMeDSe and DMeSe, respectively). Both compounds exhibited a linear signal-concentration relationship in the investigated concentration range of 0.1-1 μM with a precision on the determinations better than 3%. The method was applied for analysis of samples from cancer cell lines incubated with methylseleninic acid, selenomethionine, Se-methylselenocysteine, and sodium selenite. DMeDSe were detected in some samples. The method offers a simple and fast analysis of DMeDSe and DMeSe using standard liquid chromatography coupled with ICP-MS equipment and interfacing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328502
Dimethyl diselenide, 96%
C2H6Se2
41572
Dimethyl selenide, ≥99.0% (GC)
C2H6Se