EMAIL THIS PAGE TO A FRIEND
Australian dental journal
Antibiotics and endodontics.
PMID 2090089
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3000000 Tyrothricin, European Pharmacopoeia (EP) Reference Standard