EMAIL THIS PAGE TO A FRIEND
Farmaco (Societa chimica italiana : 1989)
omega-Dialkylaminoalkyl ethers of 6-(benzyl or phenyl)-1,3,3-trimethyl-2-oxabicyclo [2.2.2]octan-6-ol with platelet antiaggregating and local anesthetic activities.
PMID 2090141
Abstract
The synthesis of 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol 2 and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol 3 starting from (+)-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-one and phenylmagnesium bromide or benzylmagnesium chloride, respectively, is described. Alcohols 2 and 3 gave a series of omega-dialkylaminoalkyl ethers 4 by reaction as sodium salts with omega-chloroalkyldialkylamines in toluene solution. Some compounds 4, in particular those derived from alcohol 2, showed a strong platelet antiaggregating activity in vitro, superior to that of acetylsalicylic acid, as well as in general an appreciable local anesthetic activity and a weak sedative effect in mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
171565
Phenylmagnesium bromide solution, 3.0 M in diethyl ether
C6H5BrMg