EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Enantioseparation of DL-isocitric acid by a chiral ligand exchange CE with Ni(II)-D-quinic acid system.
PMID 20925051
Abstract
The ratio of citric acid to D-isocitric acid can be used to distinguish authentic and adulterated fruit juices. To separate DL-isocitric acid enantiomers, we used ligand exchange CE. D-Quinic acid was used as a chiral selector ligand and Mn(II), Fe(III), Co(II), Ni(II), Cu(II), and Zn(II) ions were used as the central ions of the chiral selector in the BGE. DL-Isocitric acid was found to be enantioseparated with the above metal ions except for Mn(II) ion. The optimum running conditions for the analysis of D- and L-isocitric acids along with citric acid, an isomer of isocitric acid, were found to be a BGE (pH 5.0) containing 30% ACN, 20 mM acetic acid, 20 mM NiSO(4), and 80 mM D-quinic acid. Under these conditions, DL-isocitric and citric acids in fruit juices were analyzed successfully.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1252
DL-Isocitric acid trisodium salt hydrate, ≥93%
C6H5Na3O7 · xH2O