EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Incapacitating CtBP to kill cancer.
PMID 20930508
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6000
α-Keto-γ-(methylthio)butyric acid sodium salt, ≥97%
C5H7NaO3S