EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Disruption of opioid-induced placebo responses by activation of cholecystokinin type-2 receptors.
PMID 20931328
Abstract
Placebos are known to induce analgesia through the activation of μ-opioid receptors in some circumstances, such as after morphine pre-conditioning, an effect that is blocked by opioid antagonists. On the basis of the anti-opioid action of cholecystokinin, here we tested whether the activation of the cholecystokinin type-2 receptors abolishes opioid-induced placebo responses. The activation of the cholecystokinin type-2 receptors was performed by means of the agonist pentagastrin, and placebo responses were obtained after morphine pre-conditioning in an experimental human model of pain (tourniquet technique). Opioid-induced placebo responses were completely disrupted by pentagastrin administration. In addition, a high correlation between the response to morphine and the response to placebo was found, and this correlation was completely abolished by pentagastrin. These results show that the cholecystokinin-2 receptor agonist, pentagastrin, has the same effect as the μ-opioid receptor antagonist, naloxone, on placebo analgesia induced by morphine pre-conditioning, which suggests that the balance between cholecystokinergic and opioidergic systems is crucial in placebo responsiveness in pain. These findings also suggest that cholecystokinin type-2 receptor hyperactivity might be present in placebo non-responders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1636
Pentagastrin, ≥95% (HPLC), powder
C37H49N7O9S