EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Versatile one-pot reductive alkylation of lactams/amides via amide activation: application to the concise syntheses of bioactive alkaloids (±)-bgugaine, (±)-coniine, (+)-preussin, and (-)-cassine.
PMID 20938943