EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Boehringer Ingelheim withdraws libido drug for women.
PMID 20940204
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0797 Flibanserin, ≥98% (HPLC)
C20H21F3N4O