EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Biomimetic analogs for collagen biomineralization.
PMID 20940362
Abstract
Inability of chemical phosphorylation of sodium trimetaphosphate to induce intrafibrillar mineralization of type I collagen may be due to the failure to incorporate a biomimetic analog to stabilize amorphous calcium phosphates (ACP) as nanoprecursors. This study investigated adsorption/desorption characteristics of hydrolyzed and pH-adjusted sodium trimetaphosphate (HPA-Na(3)P(3)O(9)) to collagen. Based on those results, a 5-minute treatment time with 2.8 wt% HPA-Na(3)P(3)O(9) was used in a single-layer reconstituted collagen model to confirm that both the ACP-stabilization analog and matrix phosphoprotein analog must be present for intrafibrillar mineralization. The results of that model were further validated by complete remineralization of phosphoric-acid-etched dentin treated with the matrix phosphoprotein analog and lined with a remineralizing lining composite, and with the ACP-stabilization analog supplied in simulated body fluid. An understanding of the basic processes involved in intrafibrillar mineralization of reconstituted collagen fibrils facilitates the design of novel tissue engineering materials for hard tissue repair and regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5508
Trisodium trimetaphosphate, ≥95%
Na3O9P3