EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Endocrinology and Metabolism
Induction of ketosis in rats fed low-carbohydrate, high fat diets depends on the relative abundance of dietary fat and protein.
PMID 20943751