EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational cobalt-allergic contact dermatitis resulting from polyester resin.
PMID 20946461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
444545
Cobalt(II) 2-ethylhexanoate solution, 65 wt. % in mineral spirits
C16H30CoO4