EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Involvement of breast cancer resistance protein (BCRP/ABCG2) in the secretion of danofloxacin into milk: interaction with ivermectin.
PMID 20950350
Abstract
Danofloxacin, a veterinary fluoroquinolone antimicrobial drug, is actively secreted into milk by an as yet unknown mechanism. One of the main determinants of active drug secretion into milk is the transporter (BCRP/ABCG2). The main purpose was to determine whether danofloxacin is an in vitro substrate for Bcrp1/BCRP and to assess its involvement in danofloxacin secretion into milk. In addition, the role of potential drug-drug interactions in this process was assessed using ivermectin. Danofloxacin was transported in vitro by Bcrp1/BCRP, and ivermectin efficiently blocked this transport. Experiments with Bcrp1(-/-) mice showed no evidence of the involvement of Bcrp1 in plasma pharmacokinetics of danofloxacin. However, the milk concentration and milk-to-plasma ratio of danofloxacin were almost twofold higher in wild-type compared with Bcrp1(-/-) mice. The in vivo interaction with ivermectin was studied in sheep after co-administration of danofloxacin (1.25 mg/kg, i.m.) and ivermectin (0.2 mg/kg, s.c.). Ivermectin had no significant effect on the plasma levels of danofloxacin but significantly decreased danofloxacin concentrations in milk by almost 40%. Concomitant administration of multiple drugs, often used in veterinary therapy, may not only affect their pharmacological activity but also their secretion into milk, because of potential drug-drug interactions mediated by BCRP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33700
Danofloxacin, VETRANAL, analytical standard
C19H20FN3O3