EMAIL THIS PAGE TO A FRIEND
Genes & development
Sept4/ARTS is required for stem cell apoptosis and tumor suppression.
PMID 20952537
Abstract
Inhibitor of Apoptosis Proteins (IAPs) are frequently overexpressed in tumors and have become promising targets for developing anti-cancer drugs. IAPs can be inhibited by natural antagonists, but a physiological requirement of mammalian IAP antagonists remains to be established. Here we show that deletion of the mouse Sept4 gene, which encodes the IAP antagonist ARTS, promotes tumor development. Sept4-null mice have increased numbers of hematopoietic stem and progenitor cells, elevated XIAP protein, increased resistance to cell death, and accelerated tumor development in an Eμ-Myc background. These phenotypes are partially suppressed by inactivation of XIAP. Our results suggest that apoptosis plays an important role as a frontline defense against cancer by restricting the number of normal stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV02014
Anti-SEPT4 antibody produced in rabbit, affinity isolated antibody