EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Role of the P2Y12 receptor in the modulation of murine dendritic cell function by ADP.
PMID 20952684
Abstract
The effects of ADP on the biology of dendritic cells have been studied much less than those of ATP or adenosine. In this study, we showed that adenosine-5'-O-(2-thiodiphosphate) (ADPβS) induced intracellular Ca(2+) transients in murine dendritic cells (DCs). This effect was abolished by AR-C69931MX, a dual P2Y(12) and P2Y(13) receptor antagonist. RT-PCR experiments revealed the expression of both P2Y(12) and P2Y(13) mRNA in DCs. The Ca(2+) response to ADPβS was maintained in P2Y(13)-deficient DCs, whereas it was abolished completely in P2Y(12)(-/-) DCs. ADPβS stimulated FITC-dextran and OVA capture in murine DCs through macropinocytosis, and this effect was abolished in P2Y(12)(-/-) DCs. ADPβS had a similar effect on FITC-dextran uptake by human monocyte-derived DCs. OVA loading in the presence of ADPβS increased the capacity of DCs to stimulate OVA-specific T cells, whereas ADPβS had no effect on the ability of DCs to stimulate allogeneic T cells. Moreover, after immunization against OVA, the serum level of anti-OVA IgG1 was significantly lower in P2Y(12)(-/-) mice than that in wild-type controls. In conclusion, we have shown that the P2Y(12) receptor is expressed in murine DCs and that its activation increased Ag endocytosis by DCs with subsequent enhancement of specific T cell activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8016
Adenosine 5′-[β-thio]diphosphate trilithium salt, ≥80% (HPLC)
C10H12Li3N5O9P2S