EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Electrochemical behavior and voltammetric determination of 2,4,6-triaminopyrimidine at glassy carbon electrode modified with multi-walled carbon nanotubes/nafion.
PMID 20953050
Abstract
A multi-wall carbon nanotubes (MWCNTs) composite with Nafion was modified on a glass carbon electrode. The modified electrode was then used as a voltammetric sensor in detecting 2,4,6-triaminopyrimidine (TAP). The surface morphology of the Nafion/MWCNTs composite film was characterized by atomic force microscopy (AFM), and the electrochemical behavior of TAP at this sensor was investigated in detail. The results indicated that the Nafion/MWCNTs modified electrode exhibited efficient electrocatalytic oxidation for TAP with relatively high sensitivity, stability and lifetime. Under the optimized condition using linear sweep voltammetry (LSV), the Nafion/MWCNTs modified electrode exhibited a linear voltammetric response for TAP in the concentration range of 2.0 × 10(-7) to 3.6 × 10(-5) mol L(-1), with a detection limit of 5.0 × 10(-8) mol L(-1). The electrode was applied to detect TAP added to human blood serum, with an average recovery value of 101.3%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T45802
2,4,6-Triaminopyrimidine, 97%
C4H7N5