EMAIL THIS PAGE TO A FRIEND
The Journal of general and applied microbiology
Description of Ectothiorhodospira salini sp. nov.
PMID 20953095
Abstract
Strain JA430(T) is a Gram-negative, vibrioid to spiral shaped phototrophic purple sulfur bacterium isolated from anoxic sediment of a saltern at Kanyakumari in a mineral salts medium that contained 2% NaCl (w/v). Strain JA430(T) grows optimally at 5-6% NaCl and tolerates up to 12% NaCl. Intracellular photosynthetic membranes were of the lamellar type. Bacteriochlorophyll a and carotenoids of the spirilloxanthin series are present as photosynthetic pigments. Major cellular fatty acids are C(18:1)ω7c, C(16:0), C(19:0)cycloω8c and C(16:1)ω7c/C(16:1)ω6c. Strain JA430(T) exhibits photoorganoheterotrophy and chemoorganoheterotrophy and requires para-aminobenzoic acid, pantothenate and pyridoxal phosphate for growth. Phylogenetic analysis on the basis of 16S rRNA gene sequence analysis showed that strain JA430(T) forms monophyletic group in the genus Ectothiorhodospira. The highest sequence similarity for strain JA430(T) was found with the type strains of Ectothiorhodospira variabilis DSM 21381(T) (96.1%) and Ectothiorhodospira haloalkaliphila ATCC 51935(T) (96.2%). Morphological and physiological characteristics discriminate strain JA430(T) from other species of the genus Ectothiorhodospira, for which we describe this as a novel species, Ectothiorhodospira salini sp. nov. ( = NBRC 105915(T) = KCTC 5805(T)).