EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
New method to enhance ajmalicine production in Catharanthus roseus cell cultures based on the use of cyclodextrins.
PMID 20953668
Abstract
The joint use of cyclodextrins and methyljasmonate, when accompanied by a short exposure to UV, enhanced extracellular ajmalicine accumulation to 1040 ± 26.6 mg/l in suspension cultured cells of Catharanthus roseus. The success of this strategy is due to the use of cyclodextrins, which not only induce ajmalicine biosynthesis but also promote adduct formation. This removes ajmalicine from the medium, reduces feedback inhibition and ajmalicine degradation, and allows its accumulation in the culture medium at elevated concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41111
Ajmalicine, ≥98.0% (HPLC)
C21H24N2O3