EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health. Part A
Role of metabolism in 1-bromopropane-induced hepatotoxicity in mice.
PMID 20954070
Abstract
A possible role of metabolism in 1-bromopropane (1-BP)-induced hepatotoxicity was investigated in male ICR mice. The depletion of glutathione (GSH) by formation of GSH conjugates was associated with increased hepatotoxicity in 1-BP-treated mice. The formation of S-propyl and 2-hydroxypropyl GSH conjugates were identified in the liver following 1-BP treatment. In addition, the formation of reactive metabolites of 1-BP by certain cytochrome P-450 (CYP) may be involved in 1-BP-induced hepatotoxicity. The decreased content of hepatic GSH produced by 1-BP was associated not only with increased activities of alanine aminotransferase (ALT) and aspartate aminotransferase (AST) but also with elevated levels of hepatic thiobarbituric acid-reactive substance (TBARS) in mice where metabolic enzymes were induced by pretreatment with phenobarbital. In addition, the hepatotoxicity induced by 1-BP was prevented by pretreatment with SKF-525A. Taken together, the formation of reactive metabolites by CYP and depletion of GSH may play important roles in hepatotoxicity induced by 1-BP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B78106
1-Bromopropane, 99%
C3H7Br
P1061
Proadifen hydrochloride, analytical standard, ≥95%
C23H31NO2 · HCl