EMAIL THIS PAGE TO A FRIEND
Turkiye parazitolojii dergisi
[The protoskolicidal effect of 1% polyvinylpyrrolidone-iodine (PVP-1) and 2% taurolidine on abdominal hydatidosis].
PMID 20954114
Abstract
The aim of this study is to determine the efficacy of 1% polyvinylprolidone-iodine (Betadine, PVP-I) and 2% Taurolidine as scolicidal agents for the prevention of abdominal hydatidosis defined as the rupture of the echinococcal cyst spontaneously or traumatically. The study was carried out in fifty mice randomly assigned into 5 treatment groups as following: group with no expose to any scolicidal agent, groups with 1% PVP-I for 2 and 5 minutes; groups with 2 % Taurolidine for 2 minutes, and 5 minutes. PVP-I has found to be effective according to results of staining with the eosin dye in vitro and abdominal hydatidosis in vivo, while Taurolidine was ineffective as a scolicidal agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7329
Taurolidine, >97% (NMR), powder
C7H16N4O4S2