EMAIL THIS PAGE TO A FRIEND
Radiation research
Ribonucleotide reductase inhibition enhances chemoradiosensitivity of human cervical cancers.
PMID 20954859
Abstract
For repair of damaged DNA, cells increase de novo synthesis of deoxyribonucleotide triphosphates through the rate-limiting, p53-regulated ribonucleotide reductase (RNR) enzyme. In this study we investigated whether pharmacological inhibition of RNR by 3-aminopyridine-2-carboxaldehyde thiosemicarbazone (3-AP, NSC #663249) enhanced chemoradiation sensitivity through a mechanism involving sustained DNA damage. RNR inactivation by 3-AP and resulting chemoradiosensitization were evaluated in human cervical (CaSki, C33-a) cancer cells through study of DNA damage (γ-H2AX signal) by flow cytometry, RNR subunit p53R2 and p21 protein steady-state levels by Western blot analysis and laser scanning imaging cytometry, and cell survival by colony formation assays. 3-AP treatment led to sustained radiation- and cisplatin-induced DNA damage (i.e. increased γ-H2AX signal) in both cell lines through a mechanism of inhibited RNR activity. Radiation, cisplatin and 3-AP exposure resulted in significantly elevated numbers and persistence of γ-H2AX foci that were associated with reduced clonogenic survival. DNA damage was associated with a rise in p53R2 but not p21 protein levels 6 h after treatment with radiation and/or cisplatin plus 3-AP. We conclude that blockage of RNR activity by 3-AP impairs DNA damage responses that rely on deoxyribonucleotide production and thereby may substantially increase chemoradiosensitivity of human cervical cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0568 3-AP, ≥98% (HPLC)
C7H9N5S