EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[Recurrent herpes simplex labialis infections. Possible prevention with a new therapeutic regimen].
PMID 20960953
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0035
Penciclovir
C10H15N5O3