EMAIL THIS PAGE TO A FRIEND
Circulation journal : official journal of the Japanese Circulation Society
Cytoskeletal proteins: hidden targets of cytoprotective signaling?
PMID 20962422
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31550
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%
C4H7NO2