EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Ex vivo screening of prospective peptide drugs: new approaches.
PMID 20963648
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8826
Semax, ≥98% (HPLC)
C37H51N9O10S · xC2HF3O2