EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Tetrabutylammonium bromide media aza-Michael addition of 1,2,3,6-tetrahydrophthalimide to symmetrical fumaric esters and acrylic esters under solvent-free conditions.
PMID 20966877
Abstract
The aza-Michael addition of 1,2,3,6-tetrahydrophthalimide with symmetrical fumaric esters has been performed efficiently in a solvent-free system at 100 °C and using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a base in the presence of tetrabutylammonium bromide (TBAB). The products were obtained in good to high yields within 2.5-7.0 h. This reaction worked well on linear alkyl fumarates and was not effective with nonlinear alkyl fumarates. Although the reaction was also applicable to acrylates such as n-butyl acrylate, methacrylates and crotonates were not suitable Michael acceptors for this reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N11400
Captan, analytical standard
C9H8Cl3NO2S
32054
Captan, PESTANAL®, analytical standard
C9H8Cl3NO2S