EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Surface functionalization of porous coordination nanocages via click chemistry and their application in drug delivery.
PMID 20972982
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
767360
5-Ethynyl-1,3-benzenedicarboxylic acid, 99% (HPLC)
C10H6O4