EMAIL THIS PAGE TO A FRIEND
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging
Pilot evaluation of anti-1-amino-2-[18F] fluorocyclopentane-1-carboxylic acid (anti-2-[18F] FACPC) PET-CT in recurrent prostate carcinoma.
PMID 20976627
Abstract
Anti-1-amino-2-[(18)F]fluorocyclopentane-1-carboxylic acid (anti-2-[(18)F]FACPC) is an unnatural alicyclic amino acid radiotracer with high uptake in the DU-145 prostate cancer cell line in vitro. Our goal was to determine if anti-2-[(18)F]FACPC is useful in the detection of prostate carcinoma. Five patients with elevated PSA (1.1-20.5 ng/mL) after curative therapy for prostate carcinoma underwent 60 min dynamic positron emission tomography (PET) of the pelvis after IV injection of 193-340 MBq of anti-2-[(18)F]FACPC. Uptake was compared against PET scans in the same patients with the leucine analog, anti-1-amino-3-[(18)F]fluorocyclobutane-1-carboxylic acid (anti-[(18)F]FACBC), at similar time points and validated via pathology, clinical, and imaging follow-up. At 5 min, average (±SD) SUVmax of malignant lesions is 4.1(±1.3) for anti-2-[(18)F] FACPC and 4.3(±1.1) for anti-[(18)F]FACBC. Yet, blood pool activity at 5 min is significantly higher for anti-2-[(18)F]FACPC with average (±SD) lesion/blood pool SUVmax/SUVmean ratio of 1.4 (±0.5) vs. 3.0 (±0.9) for anti-[(18)F]FACBC. At 20 min, average (±SD) SUVmax of malignant lesions is 2.6 (±1.0) for anti-2-[(18)F]FACPC and 3.4 (±0.8) for anti-[(18)F]FACBC. Yet, bladder activity at 20 min is significantly more intense for anti-2-[(18)F] FACPC with average (±SD) lesion/bladder SUVmax/SUVmean ratio of 0.3 (±0.8) vs. 2.3 (±1.4) for anti-[(18)F]FACBC. While prostate bed lesions are visible on early imaging with anti-2-[(18)F]FACPC, there is high blood pool activity obscuring nodes. As blood pool fades, nodal uptake decreases and high bladder activity then obscures pelvic structures. Compared with anti-[(18)F]FACBC, imaging characteristics for anti-2-[(18)F]FACPC are unfavorable for pelvic recurrent prostate carcinoma detection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A48105
Cycloleucine, 97%
C6H11NO2