EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Nucleotide recognition and phosphate linkage hydrolysis at a lipid cubic interface.
PMID 20977215
Abstract
Mononucleotides, when entrapped within a mono-olein-based cubic Ia3d liquid crystalline phase, have been found to undergo hydrolysis at the sugar-phosphate ester bond in spite of their natural inertness toward hydrolysis. Here, kinetics of the hydrolysis reaction and interactions between the lipid matrix and the mononucleotide adenosine 5'-monophosphate disodium salt (AMP) and its 2'-deoxy derivative (dAMP) are thoroughly investigated in order to shed some light on the mechanism of the nucleotide recognition and phosphate ester hydrolysis. Experiments evidenced that molecular recognition occurs essentially through the sn-2 and the sn-3 alcoholic OH groups of mono-olein. As deduced from the apparent activation energies, the mechanism underlying the hydrolysis reaction is the same for AMP and dAMP. Nevertheless, the reaction proceeds slower for the latter, highlighting a substantial difference in the chemical behavior of the two nucleotides. A model that explains the hydrolysis reaction is presented. Remarkably, the hydrolysis mechanism appears to be highly specific for the Ia3d phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6375
2′-Deoxyadenosine 5′-monophosphate, Sigma Grade, 98-100%
C10H14N5O6P