EMAIL THIS PAGE TO A FRIEND
Gastroenterology
D-glucose acts via sodium/glucose cotransporter 1 to increase NHE3 in mouse jejunal brush border by a Na+/H+ exchange regulatory factor 2-dependent process.
PMID 20977906
Abstract
Oral rehydration solutions reduce diarrhea-associated mortality. Stimulated sodium absorption by these solutions is mediated by the Na(+)/H(+) hydrogen exchanger NHE3 and is increased by Na(+)-glucose co-transport in vitro, but the mechanisms of this up-regulated process are only partially understood. Intracellular pH was measured in jejunal enterocytes of wild-type mice and mice with disrupted Na+/H+ exchange regulatory co-factor 2 (NHERF2-/- mice) by multiphoton microscopy. Diarrhea was induced by cholera toxin. Caco-2BBe cells that express NHE3 and the sodium/glucose cotransporter 1 (SGLT1) were studied by fluorometry, before and after siRNA-mediated knockdown of NHERF1 or NHERF2. NHE3 distribution was assessed by cell-surface biotinylation and confocal microscopy. Brush-border mobility was determined by fluorescence recovery after photobleaching and confocal microscopy. The nonmetabolized SGLT1 substrate α-methyl-D-Glu (α-MD-G) activated jejunal NHE3; this process required Akt and NHERF2. α-MD-G normalized NHE3 activity after cholera toxin-induced diarrhea. α-MD-G-stimulated jejunal NHE3 activity was defective in NHERF2-/- mice and cells with NHERF2 knockdown, but occurred normally with NHERF1 knockdown; was associated with increased NHE3 surface expression in Caco-2 cells, which also was NHERF2-dependent; was associated with dissociation of NHE3 from NHERF2 and an increase in the NHE3 mobile fraction from the brush border; and was accompanied by a NHERF2 ezrin-radixin-moesin-binding domain-dependent increase in co-precipitation of ezrin with NHE3. SGLT1-mediated Na-glucose co-transport stimulates NHE3 activity in vivo by an Akt- and NHERF2-dependent signaling pathway. It is associated with increased brush-border NHE3 and association between ezrin and NHE3. Activation of NHE3 corrects cholera toxin-induced defects in Na absorption and might contribute to the efficacy of oral rehydration solutions.